2-Bromo-5-(tert-butyl)aniline ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-tertbutylaniline

19949-25G 19949-5G
76897-748EA 198.39 USD
76897-748 76897-750
2-Bromo-5-(tert-butyl)aniline ≥97%
2-Bromo-5-(tert-butyl)aniline
Formula: C₁₀H₁₄BrN
MW: 228.12 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846050
CAS Number: 58164-14-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR