2-Bromo-3-fluorobenzeneboronic acid 95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-fluorophenylboronic acid, (2-Bromo-3-fluorophenyl)boronic acid

19945-1G 19945-500MG 19945-5G 19945-25G
76897-736EA 587.74 USD
76897-736 76897-738 76897-740 77654-084
2-Bromo-3-fluorobenzeneboronic acid 95%
2-Bromo-3-fluorobenzeneboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BBrFO₂
MW: 218.816 g/mol
Storage Temperature: Ambient
MDL Number: MFCD27996941
CAS Number: 731817-89-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR