2-Bromo-6-chloroanisole ≥95%

Supplier: AOBChem USA

Warning

19934-1G 19934-25G 19934-5G
76897-724EA 64.72 USD
76897-724 76897-726 76897-728
2-Bromo-6-chloroanisole ≥95%
2-Bromo-6-chloroanisole

Precaución: for Research Use Only

Formula: C₇H₆BrClO
Storage Temperature: Refrigerator
MDL Number: MFCD15143586
CAS Number: 174913-10-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR