Methyl 2-bromo-4-(trifluoromethyl)benzoate ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4-(trifluoromethyl)benzoic acid methyl ester

19870-25G 19870-5G
76897-694EA 240.83 USD
76897-694 76897-696
Methyl 2-bromo-4-(trifluoromethyl)benzoate ≥97% research grade
Methyl 2-bromo-4-(trifluoromethyl)benzoate
Formula: C₉H₆BrF₃O₂
Storage Temperature: Ambient
MDL Number: MFCD14697995
CAS Number: 1214334-90-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR