Methyl 2-chloro-3-fluorobenzoate ≥97%

Supplier: AOBChem USA

Warning

19802-1G 19802-25G 19802-500MG 19802-5G
76897-670EA 123.07 USD
76897-670 76897-672 76897-674 76897-676
Methyl 2-chloro-3-fluorobenzoate ≥97%
Methyl 2-chloro-3-fluorobenzoate

Precaución: for Research Use Only

Formula: C₈H₆ClFO₂
Storage Temperature: Refrigerator
MDL Number: MFCD10566821
CAS Number: 647020-70-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR