2-Methyl-4-(trifluoromethyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19716-1G 19716-25G 19716-5G
76897-656EA 62.6 USD
76897-656 76897-658 76897-660
2-Methyl-4-(trifluoromethyl)phenylboronic acid ≥97%
2-Methyl-4-(trifluoromethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BF₃O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08689550
CAS Number: 957034-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR