2-Bromo-4-fluorothioanisole ≥97%

Supplier: AOBChem USA

Warning

Synonyms: (2-Bromo-4-fluorophenyl)(methyl)sulphane

19693-1G 19693-25G 19693-5G
76897-642EA 62.6 USD
76897-642 76897-644 76897-646
2-Bromo-4-fluorothioanisole ≥97%
2-Bromo-4-fluorothioanisole

Precaución: for Research Use Only

Formula: C₇H₆BrFS
Storage Temperature: Refrigerator
MDL Number: MFCD09801042
CAS Number: 91524-70-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR