2-Methoxy-5-(methylaminocarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19650-1G 19650-25G 19650-500MG 19650-5G
76897-628EA 260.99 USD
76897-628 76897-630 76897-632 76897-634
2-Methoxy-5-(methylaminocarbonyl)phenylboronic acid ≥97%
2-Methoxy-5-(methylaminocarbonyl)phenylboronic acid
Formula: C₉H₁₂BNO₄
MW: 209.007 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22056743
CAS Number: 1137339-95-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR