2,6-Dichloro-4-formylphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

19518-1G 19518-25G 19518-500MG 19518-5G
76897-602EA 213.25 USD
76897-602 76897-604 76897-606 76897-608
2,6-Dichloro-4-formylphenylboronic acid ≥97%
2,6-Dichloro-4-formylphenylboronic acid
Formula: C₇H₅BCl₂O₃
MW: 218.83 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16295271
CAS Number: 1451392-98-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR