2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester ≥97%

Supplier: AOBChem USA

Warning

19492-1G 19492-25G 19492-500MG 19492-5G
76897-584EA 124.13 USD
76897-584 76897-586 76897-588 76897-590
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester ≥97%
2-Fluoro-4-hydroxybenzeneboronic acid pinacol ester
Formula: C₁₂H₁₆BFO₃
MW: 238.065 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16994421
CAS Number: 1029439-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR