2-Bromo-4-fluoro-5-methylaniline ≥97%

Supplier: AOBChem USA

Warning

19470-25G 19470-5G
76897-574EA 396.78 USD
76897-574 76897-576
2-Bromo-4-fluoro-5-methylaniline ≥97%
2-Bromo-4-fluoro-5-methylaniline

Precaución: for Research Use Only

Formula: C₇H₇BrFN
MW: 204.04 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11042843
CAS Number: 1065076-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR