2,4-Difluoro-3-methylbenzaldehyde ≥97%
Supplier: AOBChem USA
|
Warning
|
19390-25G
19390-5G
76897-562EA
488.02
USD
76897-562
76897-564
2,4-Difluoro-3-methylbenzaldehyde ≥97%
2,4-Difluoro-3-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆F₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD07777152 CAS Number: 847502-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...