2-Bromo-5-(1-hydroxy-1-methylethyl)pyridine ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-(6-Bromopyridin-3-yl)propan-2-ol

19272-1G 19272-25G 19272-5G
76897-536EA 72.15 USD
76897-536 76897-538 76897-540
2-Bromo-5-(1-hydroxy-1-methylethyl)pyridine ≥95%
2-Bromo-5-(1-hydroxy-1-methylethyl)pyridine
Formula: C₈H₁₀BrNO
MW: 216.075 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16988537
CAS Number: 477252-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR