4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Methyl-5-cyanophenylboronic acid pinacol ester

19226-1G 19226-25G 19226-500MG 19226-5G
76897-522EA 183.54 USD
76897-522 76897-524 76897-526 76897-528
4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile ≥97%
4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
Formula: C₁₄H₁₈BNO₂
MW: 243.10 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11617918
CAS Number: 863868-32-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR