2-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-imidazole ≥97%

Supplier: AOBChem USA

Warning

19161-1G 19161-25G 19161-500MG 19161-5G
76897-486EA 152.77 USD
76897-486 76897-488 76897-490 76897-492
2-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-imidazole ≥97%
2-Bromo-1-(tetrahydro-2H-pyran-2-yl)-1H-imidazole
Formula: C₈H₁₁BrN₂O
Storage Temperature: Refrigerator
MDL Number: MFCD11100954
CAS Number: 1065483-60-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR