2-Chloro-5-fluoro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

19142-25G 19142-5G
76897-474EA 254.62 USD
76897-474 76897-476
2-Chloro-5-fluoro-4-methylpyridine ≥97%
2-Chloro-5-fluoro-4-methylpyridine
Formula: C₆H₅ClFN
MW: 145.5600 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09864695
CAS Number: 881891-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR