2-Chloro-3-fluoro-4-hydroxypyridine ≥97%
Supplier: AOBChem USA
|
Warning
|
19073-1G
19073-25G
19073-5G
76897-454EA
80.63
USD
76897-454
76897-456
76897-458
2-Chloro-3-fluoro-4-hydroxypyridine ≥97%
2-Chloro-3-fluoro-4-hydroxypyridine
Formula:
C₅H₃ClFNO Storage Temperature: Refrigerator |
MDL Number:
MFCD13189885 CAS Number: 1184172-46-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...