6-Chloro-4-methylnicotinic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-4-methyl-3-pyridinecarboxylic acid, 6-Chloro-4-methylpyridine-3-carboxylic acid, 5-Carboxy-2-chloro-4-methylpyridine, 2-Chloro-4-methylpyridine-5-carboxylic acid

19053-25G 19053-5G
76897-436EA 265.23 USD
76897-436 76897-438
6-Chloro-4-methylnicotinic acid ≥97%
6-Chloro-4-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆ClNO₂
MW: 171.581 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12755871
CAS Number: 503555-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR