2-Fluoro-3-hydroxy-5-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Fluoro-5-hydroxy-3-picoline, 2-Fluoro-5-methylpyridin-3-ol

18987-1G 18987-25G 18987-500MG 18987-5G
76897-416EA 137.92 USD
76897-416 76897-418 76897-420 76897-422
2-Fluoro-3-hydroxy-5-methylpyridine ≥97%
2-Fluoro-3-hydroxy-5-methylpyridine
Formula: C₆H₆FNO
MW: 127.116 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD16611070
CAS Number: 1184172-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR