2,6-Difluoro-3-hydroxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

18973-1G 18973-25G 18973-500MG 18973-5G
76897-404EA 142.17 USD
76897-404 76897-406 76897-408 76897-410
2,6-Difluoro-3-hydroxybenzaldehyde ≥97%
2,6-Difluoro-3-hydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD08275284
CAS Number: 152434-88-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR