(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Methoxy-6-trifluoromethylphenylboronic acid

18924-1G 18924-25G 18924-500MG 18924-5G
76897-392EA 266.29 USD
76897-392 76897-394 76897-396 76897-398
(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid ≥97% research grade
(2-Methoxy-6-(trifluoromethyl)phenyl)boronic acid
Formula: C₈H₈BF₃O₃
Storage Temperature: Refrigerator
MDL Number: MFCD08059516
CAS Number: 1310384-19-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR