6-Fluoro-4-methylnicotinic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 5-Carboxy-2-fluoro-4-methylpyridine, 6-Fluoro-4-methylpyridine-3-carboxylic acid, 2-Fluoro-4-methylpyridine-5-carboxylic acid

18892-25G 18892-5G
76897-384EA 381.93 USD
76897-384 76897-386
6-Fluoro-4-methylnicotinic acid ≥95%
6-Fluoro-4-methylnicotinic acid

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.126 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09260452
CAS Number: 944582-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR