2-Chloro-3-methylbenzotrifluoride ≥97%

Supplier: AOBChem USA

Warning

18879-1G 18879-25G 18879-5G
76897-370EA 62.6 USD
76897-370 76897-372 76897-374
2-Chloro-3-methylbenzotrifluoride ≥97%
2-Chloro-3-methylbenzotrifluoride
Formula: C₈H₆ClF₃
Storage Temperature: Ambient
MDL Number: MFCD11226661
CAS Number: 74483-48-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR