2-Isopropoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Isopropoxypyridine-3-boronic acid, (2-Isopropoxypyridin-3-yl)boronic acid

18842-25G 18842-5G
76897-342EA 212.18 USD
76897-342 76897-344
2-Isopropoxy-3-pyridineboronic acid ≥97%
2-Isopropoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₃
MW: 181.0000 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11855840
CAS Number: 1150114-42-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR