2-Bromo-5-hydroxy-3-methylpyridine ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-5-hydroxy-3-picoline

18830-25G 18830-5G
76897-330EA 445.58 USD
76897-330 76897-332
2-Bromo-5-hydroxy-3-methylpyridine ≥97% research grade
2-Bromo-5-hydroxy-3-methylpyridine
Formula: C₆H₆BrNO
MW: 188.024 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09839267
CAS Number: 1003711-43-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR