2-Fluoro-5-methylnicotinic acid ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2-Fluoro-5-methylpyridine-3-carbxylic acid

18723-1G 18723-25G 18723-500MG 18723-5G
76897-300EA 215.37 USD
76897-300 76897-302 76897-304 76897-306
2-Fluoro-5-methylnicotinic acid ≥97% research grade
2-Fluoro-5-methylnicotinic acid
Formula: C₇H₆FNO₂
Storage Temperature: Refrigerator
MDL Number: MFCD11848712
CAS Number: 1042986-00-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR