2-Chloro-3,5-dimethylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-3,5-lutidine

18705-1G 18705-25G 18705-500MG 18705-5G
76897-284EA 130.5 USD
76897-284 76897-286 76897-288 76897-290
2-Chloro-3,5-dimethylpyridine ≥97%
2-Chloro-3,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₈ClN
MW: 141.598 g/mol
Storage Temperature: Ambient
MDL Number: MFCD12400986
CAS Number: 72093-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR