(4-Ethoxy-2,6-dimethylphenyl)boronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dimethyl-4-ethoxyphenylboronic acid

18527-25G 18527-5G
76897-240EA 418 USD
76897-240 76897-242
(4-Ethoxy-2,6-dimethylphenyl)boronic acid ≥97%
(4-Ethoxy-2,6-dimethylphenyl)boronic acid
Formula: C₁₀H₁₅BO₃
MW: 194.03 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD12026736
CAS Number: 1315342-15-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR