2-Borono-5-methoxybenzoic acid ≥97%

Supplier: AOBChem USA

Warning

18525-1G 18525-25G 18525-500MG 18525-5G
76897-228EA 379.81 USD
76897-228 76897-230 76897-232 76897-234
2-Borono-5-methoxybenzoic acid ≥97%
2-Borono-5-methoxybenzoic acid
Formula: C₈H₉BO₅
MW: 195.965 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD06203260
CAS Number: 1400976-19-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR