2-[(Diethylamino)methyl]phenylboronic acid ≥97% research grade

Supplier: AOBChem USA

Warning

18479-1G 18479-25G 18479-500MG 18479-5G
76897-204EA 129.43 USD
76897-204 76897-206 76897-208 76897-210
2-[(Diethylamino)methyl]phenylboronic acid ≥97% research grade
2-[(Diethylamino)methyl]phenylboronic acid
Formula: C₁₁H₁₈BNO₂
Storage Temperature: Ambient
MDL Number: MFCD06801713
CAS Number: 95753-24-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR