2-Chloro-4-methyl-5-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

18451-25G 18451-5G
76897-192EA 212.18 USD
76897-192 76897-194
2-Chloro-4-methyl-5-pyridineboronic acid ≥97%
2-Chloro-4-methyl-5-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₂
Storage Temperature: Ambient
MDL Number: MFCD07368882
CAS Number: 913836-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR