2-Fluoro-5-methylpyridine-3-carbaldehyde ≥97%

Supplier: AOBChem USA

Warning

18355-1G 18355-25G 18355-500MG 18355-5G
76897-170EA 201.58 USD
76897-170 76897-172 76897-174 76897-176
2-Fluoro-5-methylpyridine-3-carbaldehyde ≥97%
2-Fluoro-5-methylpyridine-3-carbaldehyde
Formula: C₇H₆FNO
Storage Temperature: Refrigerator
MDL Number: MFCD12025467
CAS Number: 1160993-95-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR