2-Chloro-5-hydroxy-3-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 6-Chloro-5-methyl-3-pyridinol, 6-Chloro-5-methylpyridin-3-ol

18310-1G 18310-25G 18310-5G
76897-156EA 62.6 USD
76897-156 76897-158 76897-160
2-Chloro-5-hydroxy-3-methylpyridine ≥97%
2-Chloro-5-hydroxy-3-methylpyridine
Formula: C₆H₆ClNO
MW: 143.57 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11519295
CAS Number: 54232-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR