2,6-Dibromo-3-pyridinecarboxylic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2,6-Dibromopyridine-3-carboxylic acid

18278-25G 18278-5G
76897-148EA 262.05 USD
76897-148 76897-150
2,6-Dibromo-3-pyridinecarboxylic acid ≥97%
2,6-Dibromo-3-pyridinecarboxylic acid
Formula: C₆H₃Br₂NO₂
MW: 280.90 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11846496
CAS Number: 55304-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR