2-Isopropoxypyridine ≥97%

Supplier: AOBChem USA

Warning

18179-25G 18179-5G
76897-138EA 375.56 USD
76897-138 76897-140
2-Isopropoxypyridine ≥97%
2-Isopropoxypyridine

Precaución: for Research Use Only

Formula: C₈H₁₁NO
Storage Temperature: Ambient
MDL Number: MFCD11113404
CAS Number: 16096-13-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR