2-Bromo-3,6-difluorobenzoic acid ≥95%

Supplier: AOBChem USA

Warning

18133-1G 18133-25G 18133-5G
76897-128EA 91.24 USD
76897-128 76897-130 76897-132
2-Bromo-3,6-difluorobenzoic acid ≥95%
2-Bromo-3,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃BrF₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD11849930
CAS Number: 124244-65-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR