2-Methyl-5-pyrimidineboronic acid ≥97%

Supplier: AOBChem USA

Warning

18037-1G 18037-25G 18037-5G
76897-106EA 73.21 USD
76897-106 76897-108 76897-110
2-Methyl-5-pyrimidineboronic acid ≥97%
2-Methyl-5-pyrimidineboronic acid

Precaución: for Research Use Only

Formula: C₅H₇BN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD07375144
CAS Number: 1034924-06-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR