2-Isoproxy-3-methylpyridine-5-boronic acid ≥97%

Supplier: AOBChem USA

Warning

18001-1G 18001-25G 18001-5G
76897-090EA 64.72 USD
76897-090 76897-092 76897-094
2-Isoproxy-3-methylpyridine-5-boronic acid ≥97%
2-Isoproxy-3-methylpyridine-5-boronic acid

Precaución: for Research Use Only

Formula: C₉H₁₄BNO₃
Storage Temperature: Refrigerator
MDL Number: MFCD13181629
CAS Number: 1402238-33-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR