2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1,3,5-triifluorobenzene ≥97%

Supplier: AOBChem USA

Warning

17997-1G 17997-25G 17997-5G 17997-500MG
76897-080EA 66.84 USD
76897-080 76897-082 76897-084 77649-020
2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1,3,5-triifluorobenzene ≥97%
2-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1,3,5-triifluorobenzene
Formula: C₁₂H₁₇F₃OSi
MW: 262.343 g/mol
Storage Temperature: Ambient
MDL Number: MFCD32902557
CAS Number: 2173015-27-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR