2-Bromo-4,5-dimethylaniline ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-4,5-dimethyl aniline

17966-1G 17966-25G 17966-5G
76897-066EA 64.72 USD
76897-066 76897-068 76897-070
2-Bromo-4,5-dimethylaniline ≥95%
2-Bromo-4,5-dimethylaniline

Precaución: for Research Use Only

Formula: C₈H₁₀BrN
MW: 200.076 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00031416
CAS Number: 22364-29-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR