2-Chloro-6-methoxy-3-pyridineboronic acid ≥97%

Supplier: AOBChem USA

Warning

Synonyms: 2-Chloro-6-methoxypyridine-3-lboronic acid

17942-1G 17942-25G 17942-500MG 17942-5G
76897-054EA 142.17 USD
76897-054 76897-056 76897-058 76897-060
2-Chloro-6-methoxy-3-pyridineboronic acid ≥97%
2-Chloro-6-methoxy-3-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₆H₇BClNO₃
MW: 187.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11053826
CAS Number: 1072946-25-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR