2,5-Dibromopyridine-4-carboxaldehyde ≥97% research grade

Supplier: AOBChem USA

Warning

Synonyms: 2,5-Dibromoisonicotinaldehyde, 2,5-Dibromo-pyridine-4-carbaldehyde

17905-1G 17905-25G 17905-5G
76897-044EA 62.6 USD
76897-044 76897-046 76897-048
2,5-Dibromopyridine-4-carboxaldehyde ≥97% research grade
2,5-Dibromopyridine-4-carboxaldehyde
Formula: C₆H₃Br₂NO
Storage Temperature: Refrigerator
MDL Number: MFCD10574709
CAS Number: 959244-28-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR