2-Bromo-3-chloro-4-pyridineboronic acid ≥95%

Supplier: AOBChem USA

Warning

Synonyms: 2-Bromo-3-chloropyridine-4-boronic acid

17825-1G 17825-25G 17825-5G
76897-004EA 62.6 USD
76897-004 76897-006 76897-008
2-Bromo-3-chloro-4-pyridineboronic acid ≥95%
2-Bromo-3-chloro-4-pyridineboronic acid

Precaución: for Research Use Only

Formula: C₅H₄BBrClNO₂
MW: 236.26 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08690231
CAS Number: 1003043-31-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR