2-Fluoro-3-hydroxy-4-methoxybenzaldehyde ≥97%

Supplier: AOBChem USA

Warning

17761-1G 17761-25G 17761-500MG 17761-5G
76896-984EA 203.7 USD
76896-984 76896-986 76896-988 76896-990
2-Fluoro-3-hydroxy-4-methoxybenzaldehyde ≥97%
2-Fluoro-3-hydroxy-4-methoxybenzaldehyde
Formula: C₈H₇FO₃
MW: 170.138 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08669884
CAS Number: 79418-73-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR