2-Bromo-5-chloro-4-methylpyridine ≥97%

Supplier: AOBChem USA

Warning

17740-25G 17740-5G
76896-968EA 595.17 USD
76896-968 76896-970
2-Bromo-5-chloro-4-methylpyridine ≥97%
2-Bromo-5-chloro-4-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₅BrClN
Storage Temperature: Ambient
MDL Number: MFCD11846275
CAS Number: 885267-40-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR