2-Fluoro-4-(N-ethylaminocarbonyl)phenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17685-1G 17685-25G 17685-500MG 17685-5G
76896-930EA 171.87 USD
76896-930 76896-932 76896-934 76896-936
2-Fluoro-4-(N-ethylaminocarbonyl)phenylboronic acid ≥97%
2-Fluoro-4-(N-ethylaminocarbonyl)phenylboronic acid
Formula: C₉H₁₁BFNO₃
MW: 210.998 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11856044
CAS Number: 874289-29-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR