2-(2-Bromo-4-fluorophenoxy)propane ≥95%

Supplier: AOBChem USA

Warning

17630-1G 17630-25G 17630-5G
76896-908EA 62.6 USD
76896-908 76896-910 76896-912
2-(2-Bromo-4-fluorophenoxy)propane ≥95%
2-(2-Bromo-4-fluorophenoxy)propane
Formula: C₉H₁₀BrFO
MW: 233.078 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070761
CAS Number: 202865-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR