2,5-Dibromobenzonitrile ≥97%

Supplier: AOBChem USA

Warning

17592-25G 17592-5G
76896-902EA 453.01 USD
76896-902 76896-904
2,5-Dibromobenzonitrile ≥97%
2,5-Dibromobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃Br₂N
MW: 260.9134 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00672994
CAS Number: 57381-41-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR