2,6-Dichloro-3-nitrophenylboronic acid ≥97%

Supplier: AOBChem USA

Warning

17532-1G 17532-25G 17532-5G
76896-880EA 102.91 USD
76896-880 76896-882 76896-884
2,6-Dichloro-3-nitrophenylboronic acid ≥97%
2,6-Dichloro-3-nitrophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₄BCl₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD10699640
CAS Number: 1072946-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR