2,5-Dibromo-4-iodopyridine ≥95% research grade

Supplier: AOBChem USA

Warning

17413-25G 17413-5G
76896-832EA 381.93 USD
76896-832 76896-834
2,5-Dibromo-4-iodopyridine ≥95% research grade
2,5-Dibromo-4-iodopyridine
Formula: C₅H₂Br₂IN
MW: 362.79 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13181646
CAS Number: 1061357-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR